Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGTGCTCCGGCTAAAGGCTTTG[C/T]CACATTCATTGCACTTATAGGGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602277 | ||||||||||||||||||||
Literature Links: |
ZNF184 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF184 - zinc finger protein 184 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318891.1 | 2312 | Missense Mutation | GAC,GGC | D,G 676 | NP_001305820.1 | |
NM_001318892.1 | 2312 | Missense Mutation | GAC,GGC | D,G 676 | NP_001305821.1 | |
NM_001318893.1 | 2312 | Missense Mutation | GAC,GGC | D,G 600 | NP_001305822.1 | |
NM_007149.2 | 2312 | Missense Mutation | GAC,GGC | D,G 676 | NP_009080.2 | |
XM_011514861.2 | 2312 | Missense Mutation | GAC,GGC | D,G 600 | XP_011513163.1 |