Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCAGAAATGGCCTGAGAGACCC[A/T]AACACCAGGTGGACGTTCCCCATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEP1A - meprin A subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005588.2 | 255 | Silent Mutation | CCA,CCT | P,P 71 | NP_005579.2 | |
XM_011514628.1 | 255 | Silent Mutation | CCA,CCT | P,P 99 | XP_011512930.1 | |
XM_011514629.1 | 255 | Silent Mutation | CCA,CCT | P,P 71 | XP_011512931.1 |