Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGGCATGTAGTTGTCCAGGTC[A/T]TTCACCTCCCCATCACTGTAGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRR1 - gamma-aminobutyric acid type A receptor rho1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256703.1 | 1625 | Silent Mutation | AAA,AAT | K,N 390 | NP_001243632.1 | |
NM_001256704.1 | 1625 | Silent Mutation | AAA,AAT | K,N 320 | NP_001243633.1 | |
NM_001267582.1 | 1625 | Silent Mutation | AAA,AAT | K,N 320 | NP_001254511.1 | |
NM_002042.4 | 1625 | Silent Mutation | AAA,AAT | K,N 407 | NP_002033.2 | |
XM_017010689.1 | 1625 | Silent Mutation | AAA,AAT | K,N 320 | XP_016866178.1 |