Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGCGGCGTTAGGGGCGTCCTC[C/T]GGAGGCAGGGGCGCCGGCACACCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606998 MIM: 602996 | ||||||||||||||||||||
Literature Links: |
FLOT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLOT1 - flotillin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HCG20 - HLA complex group 20 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IER3 - immediate early response 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003897.3 | 365 | Silent Mutation | CCA,CCG | P,P 111 | NP_003888.2 |