Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACAGGAGGCTGGTGCATTCGCT[C/G]TTCGCCTTTCCAGACCCCAGTACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603384 | ||||||||||||||||||||
Literature Links: |
SYNGAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYNGAP1 - synaptic Ras GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB9 - zinc finger and BTB domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152735.3 | 786 | Missense Mutation | TCT,TGT | S,C 173 | NP_689948.1 |