Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCTGTGGTCCCCGTGGAAGTCC[A/G]CCTTCTTGTTGGTGTTCTTGATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606582 MIM: 612266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLL1 - delta like canonical Notch ligand 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005618.3 | 2326 | Missense Mutation | GCG,GTG | A,V 619 | NP_005609.3 | |
XM_005266934.3 | 2326 | Intron | XP_005266991.1 |
FAM120B - family with sequence similarity 120B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01624 - long intergenic non-protein coding RNA 1624 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC285804 - uncharacterized LOC285804 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |