Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGGCGCGTACTAAGCAGACGGC[C/T]CGTAAATCCACAGGCGGTAAAGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142210 MIM: 602786 MIM: 602798 MIM: 602813 | ||||||||||||||||||||
Literature Links: |
HIST1H1D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST1H1D - histone cluster 1, H1d | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2AE - histone cluster 1, H2ae | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2BG - histone cluster 1, H2bg | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3E - histone cluster 1, H3e | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003532.2 | 24 | Silent Mutation | GCC,GCT | A,A 8 | NP_003523.1 |