Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGCATGATCCTGATGACACCCG[A/G]CGGAAAAGCAGGCTGGAGCCTCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTCH1 - mitochondrial carrier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271641.1 | 1512 | Silent Mutation | CGC,CGT | R,R 378 | NP_001258570.1 | |
NM_014341.2 | 1512 | Silent Mutation | CGC,CGT | R,R 361 | NP_055156.1 | |
XM_005248978.1 | 1512 | UTR 3 | XP_005249035.1 | |||
XM_005248979.1 | 1512 | Intron | XP_005249036.1 | |||
XM_011514450.1 | 1512 | Intron | XP_011512752.1 | |||
XM_017010642.1 | 1512 | UTR 3 | XP_016866131.1 | |||
XM_017010643.1 | 1512 | Silent Mutation | CGC,CGT | R,R 222 | XP_016866132.1 |
PI16 - peptidase inhibitor 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |