Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCATCCCCAGCCTCCCCTCCCA[A/G]CTGCTGGGCTTGGCTTTCCCACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607744 MIM: 611389 | ||||||||||||||||||||
Literature Links: |
FRS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRS3 - fibroblast growth factor receptor substrate 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006653.4 | 2125 | Silent Mutation | CTG,TTG | L,L 339 | NP_006644.1 | |
XM_011514254.2 | 2125 | Silent Mutation | CTG,TTG | L,L 339 | XP_011512556.1 | |
XM_011514255.2 | 2125 | Silent Mutation | CTG,TTG | L,L 339 | XP_011512557.1 | |
XM_017010193.1 | 2125 | Silent Mutation | CTG,TTG | L,L 302 | XP_016865682.1 |
PRICKLE4 - prickle planar cell polarity protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |