Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTATCAGAGCCCATAGACCTTG[C/T]ATAGCTGCTGCCCATTCAAAACCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613361 MIM: 603571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC18B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC18B1 - solute carrier family 18 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052831.2 | 1382 | Missense Mutation | ATA,ATG | I,M 410 | NP_439896.1 | |
XM_017010236.1 | 1382 | Missense Mutation | ATA,ATG | I,M 363 | XP_016865725.1 |
VNN2 - vanin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |