Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTGTATTGTAACCAGAATGCA[C/T]AATAAATATTCTTGCCCCAGAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615353 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC28A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC28A - coiled-coil domain containing 28A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ECT2L - epithelial cell transforming 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077706.2 | Intron | NP_001071174.1 | ||||
NM_001195037.2 | Intron | NP_001181966.1 | ||||
XM_006715472.3 | Intron | XP_006715535.1 | ||||
XM_011535795.2 | Intron | XP_011534097.1 | ||||
XM_011535797.2 | Intron | XP_011534099.1 | ||||
XM_017010828.1 | Intron | XP_016866317.1 | ||||
XM_017010829.1 | Intron | XP_016866318.1 | ||||
XM_017010830.1 | Intron | XP_016866319.1 |