Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGATCTTAAGCAACCGGGTGATC[A/G]TGTTGCCATTTGCTAATATGCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTFR2 - mitochondrial fission regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099286.2 | Intron | NP_001092756.1 | ||||
NM_001318738.1 | Intron | NP_001305667.1 | ||||
NM_138419.4 | Intron | NP_612428.2 | ||||
XM_011535410.2 | Intron | XP_011533712.1 | ||||
XM_011535412.2 | Intron | XP_011533714.1 | ||||
XM_011535413.2 | Intron | XP_011533715.1 |