Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAACCCTCAAGTCACAGGCAGGT[C/T]TTAGGTTAAAGGGCGTGGTCTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNGTT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNGTT - RNA guanylyltransferase and 5'-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286426.1 | 1946 | UTR 3 | NP_001273355.1 | |||
NM_001286428.1 | 1946 | UTR 3 | NP_001273357.1 | |||
NM_003800.4 | 1946 | UTR 3 | NP_003791.3 | |||
XM_017011401.1 | 1946 | UTR 3 | XP_016866890.1 | |||
XM_017011402.1 | 1946 | Intron | XP_016866891.1 |