Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTGCATTCTTCAGCATTAACAG[C/T]GTGTAGTTTTTCTCTCTCTCTTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608024 | ||||||||||||||||||||
Literature Links: |
C6orf201 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf201 - chromosome 6 open reading frame 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085401.2 | 1015 | Intron | NP_001078870.1 | |||
XM_005249112.3 | 1015 | Intron | XP_005249169.1 | |||
XM_011514613.2 | 1015 | Intron | XP_011512915.1 | |||
XM_017010861.1 | 1015 | Intron | XP_016866350.1 |
ECI2 - enoyl-CoA delta isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166010.1 | 1015 | Missense Mutation | ACT,GCT | T,A 332 | NP_001159482.1 | |
NM_006117.2 | 1015 | Missense Mutation | ACT,GCT | T,A 332 | NP_006108.2 | |
NM_206836.2 | 1015 | Missense Mutation | ACT,GCT | T,A 362 | NP_996667.2 | |
XM_006714957.1 | 1015 | Missense Mutation | ACT,GCT | T,A 332 | XP_006715020.1 | |
XM_017010170.1 | 1015 | Missense Mutation | ACT,GCT | T,A 302 | XP_016865659.1 |