Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTGGGTTCTGATCTTCACATTG[A/C]AGTGAAGGGTTATGAGGATGGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613594 MIM: 613595 | ||||||||||||||||||||
Literature Links: |
BTN3A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTN3A3 - butyrophilin subfamily 3 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242803.1 | 630 | Intron | NP_001229732.1 | |||
NM_006994.4 | 630 | Missense Mutation | GAA,GCA | E,A 153 | NP_008925.1 | |
NM_197974.2 | 630 | Missense Mutation | GAA,GCA | E,A 111 | NP_932078.2 |