Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGTACACAAGCACGCTCTTCAC[G/T]ACGGGCTGAGACAGGTGGCTGGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605755 MIM: 608211 | ||||||||||||||||||||
Literature Links: |
DCDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCDC2 - doublecortin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195610.1 | 254 | Silent Mutation | NP_001182539.1 | |||
NM_016356.4 | 254 | Silent Mutation | NP_057440.2 |
KAAG1 - kidney associated antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181337.3 | 254 | Missense Mutation | GAC,TAC | D,Y 23 | NP_851854.1 |