Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTAACCTCCACGAGCTGCTTC[C/T]GAATCACCTGATGGAGACGCTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf201 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf201 - chromosome 6 open reading frame 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085401.2 | 411 | Missense Mutation | CCG,CTG | P,L 10 | NP_001078870.1 | |
XM_005249112.3 | 411 | Missense Mutation | CCG,CTG | P,L 10 | XP_005249169.1 | |
XM_011514613.2 | 411 | Missense Mutation | CCG,CTG | P,L 10 | XP_011512915.1 | |
XM_017010861.1 | 411 | Intron | XP_016866350.1 |
FAM217A - family with sequence similarity 217 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173563.2 | 411 | Intron | NP_775834.2 | |||
XM_005248951.3 | 411 | Intron | XP_005249008.1 | |||
XM_006715025.2 | 411 | Intron | XP_006715088.1 | |||
XM_006715026.3 | 411 | Intron | XP_006715089.1 | |||
XM_011514413.2 | 411 | Intron | XP_011512715.1 | |||
XM_011514414.2 | 411 | Intron | XP_011512716.1 | |||
XM_011514415.1 | 411 | Intron | XP_011512717.1 | |||
XM_011514416.2 | 411 | Intron | XP_011512718.1 | |||
XM_011514417.1 | 411 | Intron | XP_011512719.1 | |||
XM_017010480.1 | 411 | Intron | XP_016865969.1 | |||
XM_017010481.1 | 411 | Intron | XP_016865970.1 | |||
XM_017010482.1 | 411 | Intron | XP_016865971.1 |