Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGACCTTAGGAGGTAGAGGGG[A/G]CAATGAGGATGGAGCTGGGAGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605086 MIM: 609714 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY10P1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY10P1 - adenylate cyclase 10, soluble pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105375056 - uncharacterized LOC105375056 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TREM2 - triggering receptor expressed on myeloid cells 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TREML1 - triggering receptor expressed on myeloid cells like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271807.1 | 857 | UTR 3 | NP_001258736.1 | |||
NM_001271808.1 | 857 | Missense Mutation | CCC,TCC | P,S 155 | NP_001258737.1 | |
NM_178174.3 | 857 | Missense Mutation | CCC,TCC | P,S 266 | NP_835468.1 | |
XM_017010822.1 | 857 | Missense Mutation | CCC,TCC | P,S 325 | XP_016866311.1 | |
XM_017010823.1 | 857 | Missense Mutation | CCC,TCC | P,S 266 | XP_016866312.1 | |
XM_017010824.1 | 857 | Missense Mutation | CCC,TCC | P,S 266 | XP_016866313.1 | |
XM_017010825.1 | 857 | UTR 3 | XP_016866314.1 |