Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGTCAGCAGAGGGTGGCTTTG[A/G]TGCTGTCATCTGCCTTGGAAACAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606628 MIM: 601498 | ||||||||||||||||||||
Literature Links: |
CNPY3-GNMT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNPY3-GNMT - CNPY3-GNMT readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318856.1 | 415 | Missense Mutation | GAT,GGT | D,G 68 | NP_001305785.1 | |
NM_001318857.1 | 415 | Missense Mutation | GAT,GGT | D,G 73 | NP_001305786.1 | |
NM_001318858.1 | 415 | Missense Mutation | GAT,GGT | D,G 73 | NP_001305787.1 |
GNMT - glycine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318865.1 | 415 | Missense Mutation | GAT,GGT | D,G 134 | NP_001305794.1 | |
NM_018960.5 | 415 | Missense Mutation | GAT,GGT | D,G 134 | NP_061833.1 |
PEX6 - peroxisomal biogenesis factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |