Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACCTGATATGTTCCCAGTCAAC[A/G]CCTTCAAAAAAAGAGTTACTTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615932 MIM: 606964 | ||||||||||||||||||||
Literature Links: |
KCTD20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD20 - potassium channel tetramerization domain containing 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK38 - serine/threonine kinase 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305102.1 | 1481 | Silent Mutation | GGC,GGT | G,G 384 | NP_001292031.1 | |
NM_007271.3 | 1481 | Silent Mutation | GGC,GGT | G,G 384 | NP_009202.1 | |
XM_006714988.3 | 1481 | Silent Mutation | GGC,GGT | G,G 384 | XP_006715051.1 | |
XM_006714989.3 | 1481 | Silent Mutation | GGC,GGT | G,G 384 | XP_006715052.1 | |
XM_017010226.1 | 1481 | Silent Mutation | GGC,GGT | G,G 384 | XP_016865715.1 |