Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAGCAAGCACAGCAAAATATTC[A/G]TAACTGTCCAATACAGCCTTATCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611976 | ||||||||||||||||||||
Literature Links: |
MRPS10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPS10 - mitochondrial ribosomal protein S10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018141.3 | 320 | Silent Mutation | TAC,TAT | Y,Y 91 | NP_060611.2 | |
XM_005249206.2 | 320 | Silent Mutation | TAC,TAT | Y,Y 90 | XP_005249263.1 | |
XM_011514724.2 | 320 | Silent Mutation | TAC,TAT | Y,Y 101 | XP_011513026.1 | |
XM_011514725.2 | 320 | Silent Mutation | TAC,TAT | Y,Y 100 | XP_011513027.1 | |
XM_017010996.1 | 320 | Silent Mutation | TAC,TAT | Y,Y 50 | XP_016866485.1 |