Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTGAAAGAAGGCAGTTTTCACA[C/T]GATAAGAAGAGAATTTATCCAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606286 MIM: 613973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX43 - DEAD-box helicase 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MB21D1 - Mab-21 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138441.2 | 1459 | Missense Mutation | CAT,CGT | H,R 437 | NP_612450.2 | |
XM_017010232.1 | 1459 | Missense Mutation | CAT,CGT | H,R 437 | XP_016865721.1 |