Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATACTCTTCCTCTCTCAGCAGCC[A/G]CAGCAGGATGTTTTTTTTATTCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607043 | ||||||||||||||||||||
Literature Links: |
TRAF3IP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAF3IP2 - TRAF3 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164281.2 | 2854 | Missense Mutation | CGG,TGG | R,W 541 | NP_001157753.1 | |
NM_001164283.2 | 2854 | Missense Mutation | CGG,TGG | R,W 86 | NP_001157755.1 | |
NM_147200.2 | 2854 | Missense Mutation | CGG,TGG | R,W 551 | NP_671733.2 | |
NM_147686.3 | 2854 | Missense Mutation | CGG,TGG | R,W 542 | NP_679211.2 | |
XM_006715319.3 | 2854 | Missense Mutation | CGG,TGG | R,W 86 | XP_006715382.1 | |
XM_011535386.1 | 2854 | Missense Mutation | CGG,TGG | R,W 551 | XP_011533688.1 |
TRAF3IP2-AS1 - TRAF3IP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |