Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGCCCCACATTGCTGGACCACA[C/T]GGTAGCCGCCAGCCCATACTTAACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606467 | ||||||||||||||||||||
Literature Links: |
ALDH8A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH8A1 - aldehyde dehydrogenase 8 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193480.1 | 1186 | Missense Mutation | ATG,GTG | M,V 373 | NP_001180409.1 | |
NM_022568.3 | 1186 | Missense Mutation | ATG,GTG | M,V 423 | NP_072090.1 | |
NM_170771.2 | 1186 | Missense Mutation | ATG,GTG | M,V 369 | NP_739577.1 |