Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGATGACTGCAACATGCTGAGG[C/T]TGGCTCCGACATTTACTACAGATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602776 | ||||||||||||||||||||
Literature Links: |
REV3L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
REV3L - REV3 like, DNA directed polymerase zeta catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286431.1 | 9810 | Silent Mutation | CAA,CAG | Q,Q 2982 | NP_001273360.1 | |
NM_001286432.1 | 9810 | Silent Mutation | CAA,CAG | Q,Q 2982 | NP_001273361.1 | |
NM_002912.4 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3060 | NP_002903.3 | |
XM_011536028.1 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3087 | XP_011534330.1 | |
XM_011536029.2 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3086 | XP_011534331.1 | |
XM_011536030.2 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3061 | XP_011534332.1 | |
XM_011536031.2 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3009 | XP_011534333.1 | |
XM_011536032.2 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3009 | XP_011534334.1 | |
XM_011536036.2 | 9810 | Intron | XP_011534338.1 | |||
XM_017011152.1 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3008 | XP_016866641.1 | |
XM_017011153.1 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3008 | XP_016866642.1 | |
XM_017011154.1 | 9810 | Silent Mutation | CAA,CAG | Q,Q 3008 | XP_016866643.1 | |
XM_017011155.1 | 9810 | Intron | XP_016866644.1 |