Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGCGCCCTGGCCCAAGGAGGA[A/G]GCCCTTCAGCAACGGAGTGGGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C6orf223 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf223 - chromosome 6 open reading frame 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171992.2 | 1148 | Missense Mutation | AGC,GGC | S,G 15 | NP_001165463.1 | |
NM_001324369.1 | 1148 | UTR 5 | NP_001311298.1 | |||
NM_153246.5 | 1148 | Missense Mutation | AGC,GGC | S,G 15 | NP_694978.2 | |
XM_017010425.1 | 1148 | Missense Mutation | AGC,GGC | S,G 15 | XP_016865914.1 |
LOC101929705 - uncharacterized LOC101929705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |