Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTAAAGCCACAGAGCATGCAA[C/G]TGAATTGGCAAAAGAGAATATTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM26F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM26F - family with sequence similarity 26 member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010919.2 | 1091 | Missense Mutation | ACT,AGT | T,S 230 | NP_001010919.1 | |
NM_001276460.1 | 1091 | Missense Mutation | ACT,AGT | T,S 58 | NP_001263389.1 | |
XM_005266999.3 | 1091 | Missense Mutation | ACT,AGT | T,S 230 | XP_005267056.1 | |
XM_011535845.2 | 1091 | Missense Mutation | ACT,AGT | T,S 252 | XP_011534147.1 |