Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCAACTTCATCACTCTCTGTTT[C/T]GGTACACTGGGATTGCAGCTGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606111 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCHR2 - melanin concentrating hormone receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040179.1 | 1041 | Silent Mutation | CCA,CCG | P,P 242 | NP_001035269.1 | |
NM_032503.2 | 1041 | Silent Mutation | CCA,CCG | P,P 242 | NP_115892.2 |