Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGTGCCCTTTCTATGCAGCTA[C/T]TTCTGTGCCAAGCACGGATGGTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605701 MIM: 616976 | ||||||||||||||||||||
Literature Links: |
TRIM10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM10 - tripartite motif containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006778.3 | 2040 | UTR 3 | NP_006769.2 | |||
NM_052828.2 | 2040 | Missense Mutation | ATA,GTA | I,V 393 | NP_439893.2 | |
XM_011514221.1 | 2040 | UTR 3 | XP_011512523.1 | |||
XM_011514222.2 | 2040 | UTR 3 | XP_011512524.1 | |||
XM_011514223.2 | 2040 | UTR 3 | XP_011512525.1 | |||
XM_011514225.1 | 2040 | Missense Mutation | ATA,GTA | I,V 411 | XP_011512527.1 |
TRIM15 - tripartite motif containing 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM40 - tripartite motif containing 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |