Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATACCATAGATTAGAAGTTGTCA[A/G]TTTCAACAGTGAAACCCAGCTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606627 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAAM2 - dishevelled associated activator of morphogenesis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201427.1 | Intron | NP_001188356.1 | ||||
NM_015345.3 | Intron | NP_056160.2 | ||||
XM_006715039.3 | Intron | XP_006715102.3 | ||||
XM_006715040.3 | Intron | XP_006715103.3 | ||||
XM_006715042.2 | Intron | XP_006715105.1 | ||||
XM_006715043.2 | Intron | XP_006715106.1 | ||||
XM_006715045.3 | Intron | XP_006715108.1 | ||||
XM_006715046.3 | Intron | XP_006715109.1 | ||||
XM_017010630.1 | Intron | XP_016866119.1 |