Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTGTCTGTTGCTAAGGTCACC[A/C]TGGCTGACCGGGCTGCTGCCGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RBM24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM24 - RNA binding motif protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143941.1 | 108 | Missense Mutation | ATG,CTG | M,L 1 | NP_001137413.1 | |
NM_001143942.1 | 108 | Missense Mutation | ATG,CTG | M,L 59 | NP_001137414.1 | |
NM_153020.2 | 108 | Missense Mutation | ATG,CTG | M,L 14 | NP_694565.1 | |
XM_005248932.3 | 108 | Missense Mutation | ATG,CTG | M,L 59 | XP_005248989.1 | |
XM_011514387.2 | 108 | Missense Mutation | ATG,CTG | M,L 1 | XP_011512689.1 | |
XM_011514388.2 | 108 | Intron | XP_011512690.1 | |||
XM_011514389.2 | 108 | Intron | XP_011512691.1 |