Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATAAGCCTGGTAGCCTGGATTG[A/G]TTGTGTACCCAAACACACACTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR63 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR63 - G protein-coupled receptor 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143957.2 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | NP_001137429.1 | |
NM_030784.3 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | NP_110411.1 | |
XM_006715570.3 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_006715633.1 | |
XM_011536153.2 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_011534455.1 | |
XM_011536154.2 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_011534456.1 | |
XM_011536155.2 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_011534457.1 | |
XM_011536157.2 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_011534459.1 | |
XM_011536159.2 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_011534461.1 | |
XM_017011334.1 | 1592 | Missense Mutation | ACC,ATC | T,I 235 | XP_016866823.1 |