Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGGGGGTGCAGGTAGGCACTC[C/T]TGTACAGGCTGGTGTAGGTCACGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600830 | ||||||||||||||||||||
Literature Links: |
TRIM26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM26 - tripartite motif containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242783.1 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | NP_001229712.1 | |
NM_003449.4 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | NP_003440.1 | |
XM_005249374.2 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | XP_005249431.1 | |
XM_005249375.2 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | XP_005249432.1 | |
XM_005249376.2 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | XP_005249433.1 | |
XM_005249377.2 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | XP_005249434.1 | |
XM_005249378.2 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | XP_005249435.1 | |
XM_006715180.2 | 1155 | Missense Mutation | AAG,AGG | K,R 342 | XP_006715243.1 | |
XM_011514859.1 | 1155 | Missense Mutation | AAG,AGG | K,R 265 | XP_011513161.1 | |
XM_017011263.1 | 1155 | Missense Mutation | AAG,AGG | K,R 265 | XP_016866752.1 |