Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGCCCCTGGAGTTTCTGGGTCC[C/G]GGCCTGGCCATCCACCTGCACACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120820 MIM: 613815 MIM: 600985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4B - complement component 4B (Chido blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYP21A2 - cytochrome P450 family 21 subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNXB - tenascin XB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019105.6 | 11618 | Missense Mutation | CGG,GGG | R,G 3806 | NP_061978.6 | |
NM_032470.3 | 11618 | Missense Mutation | CGG,GGG | R,G 237 | NP_115859.2 |