Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606520 MIM: 604744 MIM: 610435 MIM: 606038 MIM: 610437 MIM: 611404 | ||||||||||||||||||||
Literature Links: |
C6orf25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf25 - chromosome 6 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDAH2 - dimethylarginine dimethylaminohydrolase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G6C - lymphocyte antigen 6 complex, locus G6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025261.2 | 363 | Intron | NP_079537.1 |
LY6G6D - lymphocyte antigen 6 complex, locus G6D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021246.2 | 363 | Silent Mutation | ACC,ACT | T,T 121 | NP_067069.2 |
LY6G6E - lymphocyte antigen 6 complex, locus G6E (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G6F - lymphocyte antigen 6 complex, locus G6F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |