Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTGTTGTCTCGAGGTTTACCA[C/T]TGCTCAAATCAAGACCAATCCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100678 MIM: 186980 | ||||||||||||||||||||
Literature Links: |
ACAT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAT2 - acetyl-CoA acetyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303253.1 | 1710 | Intron | NP_001290182.1 | |||
NM_005891.2 | 1710 | Intron | NP_005882.2 |
SNORA20 - small nucleolar RNA, H/ACA box 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA29 - small nucleolar RNA, H/ACA box 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCP1 - t-complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008897.1 | 1710 | Missense Mutation | AAT,AGT | N,S 337 | NP_001008897.1 | |
NM_030752.2 | 1710 | Missense Mutation | AAT,AGT | N,S 492 | NP_110379.2 |