Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATGTAAATCACTTTTCCTTTCT[A/G]TAAGACGATACAACTTGCGAAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611028 | ||||||||||||||||||||
Literature Links: |
TMEM30A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM30A - transmembrane protein 30A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143958.1 | 997 | Missense Mutation | ACA,ATA | T,I 240 | NP_001137430.1 | |
NM_018247.3 | 997 | Missense Mutation | ACA,ATA | T,I 276 | NP_060717.1 |