Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTACACCAGTGTGATCATAGGA[A/G]CAACTTTTTATAAACTGGAACTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615930 MIM: 609590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAHM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAHM - colon adenocarcinoma hypermethylated (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QKI - QKI, KH domain containing RNA binding | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301085.1 | Intron | NP_001288014.1 | ||||
NM_006775.2 | Intron | NP_006766.1 | ||||
NM_206853.2 | Intron | NP_996735.1 | ||||
NM_206854.2 | Intron | NP_996736.1 | ||||
NM_206855.2 | Intron | NP_996737.1 | ||||
XM_011536258.2 | Intron | XP_011534560.1 | ||||
XM_011536259.2 | Intron | XP_011534561.1 | ||||
XM_011536260.2 | Intron | XP_011534562.1 | ||||
XM_011536261.1 | Intron | XP_011534563.1 | ||||
XM_017011502.1 | Intron | XP_016866991.1 | ||||
XM_017011503.1 | Intron | XP_016866992.1 | ||||
XM_017011504.1 | Intron | XP_016866993.1 |