Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTCTCGATTCACAATCTGGGA[C/T]GACCCTGATGCTGGCTTCTTCACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604531 | ||||||||||||||||||||
Literature Links: |
NCR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCR2 - natural cytotoxicity triggering receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199509.1 | 342 | Silent Mutation | GAC,GAT | D,D 85 | NP_001186438.1 | |
NM_001199510.1 | 342 | Silent Mutation | GAC,GAT | D,D 85 | NP_001186439.1 | |
NM_004828.3 | 342 | Silent Mutation | GAC,GAT | D,D 85 | NP_004819.2 | |
XM_017011500.1 | 342 | Silent Mutation | GAC,GAT | D,D 93 | XP_016866989.1 |