Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTTCCTGTATTTGCCTTCGCC[C/T]TTTTCGAGCCTTTTGATTTTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C6orf10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf10 - chromosome 6 open reading frame 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286474.1 | 1849 | Missense Mutation | AAG,AGG | K,R 546 | NP_001273403.1 | |
NM_001286475.1 | 1849 | Missense Mutation | AAG,AGG | K,R 532 | NP_001273404.1 | |
NM_006781.4 | 1849 | Missense Mutation | AAG,AGG | K,R 548 | NP_006772.3 | |
XM_017010182.1 | 1849 | Missense Mutation | AAG,AGG | K,R 454 | XP_016865671.1 | |
XM_017010183.1 | 1849 | Intron | XP_016865672.1 | |||
XM_017010184.1 | 1849 | Intron | XP_016865673.1 | |||
XM_017010185.1 | 1849 | Missense Mutation | AAG,AGG | K,R 559 | XP_016865674.1 |
LOC101929163 - uncharacterized LOC101929163 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |