Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTCATTAGGCAAGAGAGTAGTT[C/T]GTTCCTCCTCTGTGCTGAGGATGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613361 MIM: 603571 | ||||||||||||||||||||
Literature Links: |
SLC18B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC18B1 - solute carrier family 18 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052831.2 | 1495 | Missense Mutation | CAA,CGA | Q,R 448 | NP_439896.1 | |
XM_017010236.1 | 1495 | Missense Mutation | CAA,CGA | Q,R 401 | XP_016865725.1 |
VNN2 - vanin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |