Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAGCAGCTGGGAAGCCTCATTG[A/G]AGCCCTCAAAGAGACGGATCAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIM38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM38 - tripartite motif containing 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006355.4 | 834 | Missense Mutation | GAA,GGA | E,G 82 | NP_006346.1 | |
XM_005248799.4 | 834 | Missense Mutation | GAA,GGA | E,G 82 | XP_005248856.1 | |
XM_005248800.4 | 834 | Missense Mutation | GAA,GGA | E,G 82 | XP_005248857.1 | |
XM_017010175.1 | 834 | Missense Mutation | GAA,GGA | E,G 82 | XP_016865664.1 |