Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAATTCTTCCCAAATATTTCAA[A/G]CACAATAATATGGCAAGCTTTGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 140581 | ||||||||||||||||||||
Literature Links: |
HSF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSF2 - heat shock transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135564.1 | 349 | Silent Mutation | AAA,AAG | K,K 54 | NP_001129036.1 | |
NM_001243094.1 | 349 | Silent Mutation | AAA,AAG | K,K 54 | NP_001230023.1 | |
NM_004506.3 | 349 | Silent Mutation | AAA,AAG | K,K 54 | NP_004497.1 |