Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGCCAGCGGTAGCAGTGAGTC[A/G]TCCAGGAGCATGAGGTCCAGATCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600744 | ||||||||||||||||||||
Literature Links: |
TFEB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TFEB - transcription factor EB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167827.2 | 1262 | Silent Mutation | GAC,GAT | D,D 453 | NP_001161299.2 | |
NM_001271943.1 | 1262 | Silent Mutation | GAC,GAT | D,D 354 | NP_001258872.1 | |
NM_001271944.1 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | NP_001258873.1 | |
NM_001271945.1 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | NP_001258874.1 | |
NM_007162.2 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | NP_009093.1 | |
XM_005249411.1 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | XP_005249468.1 | |
XM_005249412.1 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | XP_005249469.1 | |
XM_006715212.3 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | XP_006715275.1 | |
XM_006715213.2 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | XP_006715276.1 | |
XM_011514915.1 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | XP_011513217.1 | |
XM_011514916.2 | 1262 | Silent Mutation | GAC,GAT | D,D 439 | XP_011513218.1 |