Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACTCTCAGACCCCGGCTCTCGG[C/T]GGCCCCATTTGCCAGCCAGACTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612944 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASET2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNASET2 - ribonuclease T2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003730.4 | 1104 | Missense Mutation | NP_003721.2 | |||
XM_017011397.1 | 1104 | Intron | XP_016866886.1 | |||
XM_017011398.1 | 1104 | Intron | XP_016866887.1 | |||
XM_017011399.1 | 1104 | UTR 3 | XP_016866888.1 |