Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAAGAGCCAGCAGTACTTTCAA[C/G]AAATGACTTATAAATTTGAGAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613542 MIM: 607191 | ||||||||||||||||||||
Literature Links: |
MTRF1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTRF1L - mitochondrial translational release factor 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGS17 - regulator of G-protein signaling 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012419.4 | 754 | Missense Mutation | CTT,GTT | L,V 200 | NP_036551.3 | |
XM_017010733.1 | 754 | Intron | XP_016866222.1 |