Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGTGAATTAGAACGGGGCAGT[A/C]GTCTGGACTCCGAGTCCTGGTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611401 MIM: 602593 MIM: 613525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf15 - chromosome 6 open reading frame 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014070.2 | 530 | Missense Mutation | NP_054789.2 |
CDSN - corneodesmosin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSORS1C1 - psoriasis susceptibility 1 candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |