Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTCAGTTAACAGATTAACCTAA[G/T]AGCAATATATTAATACATTCTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611814 MIM: 616799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELOVL2 - ELOVL fatty acid elongase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017770.3 | 1202 | UTR 3 | NP_060240.3 | |||
XM_011514716.2 | 1202 | UTR 3 | XP_011513018.1 | |||
XM_011514717.2 | 1202 | UTR 3 | XP_011513019.1 | |||
XM_017010985.1 | 1202 | Intron | XP_016866474.1 |
SYCP2L - synaptonemal complex protein 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |