Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGTCATCATCAAGGCAGCGTTC[C/T]CCTGTTAAAATAGATGTTGTGTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613351 | ||||||||||||||||||||
Literature Links: |
ARHGAP18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP18 - Rho GTPase activating protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033515.2 | 1866 | Silent Mutation | GGA,GGG | G,G 634 | NP_277050.2 | |
XM_005267212.2 | 1866 | Silent Mutation | GGA,GGG | G,G 684 | XP_005267269.2 | |
XM_005267213.1 | 1866 | UTR 3 | XP_005267270.1 | |||
XM_017011494.1 | 1866 | UTR 3 | XP_016866983.1 |