Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGTTTCATAGGATAAACTCATA[A/G]AAGAGTATAAAAAGCATAAAAACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP7 - A-kinase anchoring protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004842.3 | 554 | Intron | NP_004833.1 | |||
NM_016377.3 | 554 | Intron | NP_057461.2 | |||
NM_138633.2 | 554 | Intron | NP_619539.1 | |||
XM_005267228.1 | 554 | Intron | XP_005267285.1 | |||
XM_005267229.4 | 554 | UTR 5 | XP_005267286.1 | |||
XM_006715614.2 | 554 | Intron | XP_006715677.1 | |||
XM_011536262.1 | 554 | Intron | XP_011534564.1 | |||
XM_017011506.1 | 554 | Intron | XP_016866995.1 | |||
XM_017011507.1 | 554 | Intron | XP_016866996.1 | |||
XM_017011508.1 | 554 | Intron | XP_016866997.1 | |||
XM_017011509.1 | 554 | Intron | XP_016866998.1 | |||
XM_017011510.1 | 554 | Intron | XP_016866999.1 | |||
XM_017011511.1 | 554 | Intron | XP_016867000.1 |